Please login to your online account to display your discounted pricing
Fisherbrand™ Schellbach Burets with Three-Way Stopcock
Automatic zeroing
$759.70 - $759.70
Products
Catalog Number Mfr. No. Price Quantity    
03-750C
  Accessories
fisher scientific
TG1822034
Each for $759.70
Case of 6 Each for $4,558.20
Description & Specifications
  • Tip located in overflow cup with drain at side
  • Calibrated to a tip at top
  • Black graduations with Schellbach stripe
Meet ASTM™ E 287