Fisherbrand™ Schellbach Burets with Straight Standard Taper Glass Stopcock
Manufacturer:  Fisherbrand™ 703740A
 View more versions of this product
Catalog No. 03-740A
Description
Description
  • Accentuate meniscus for easy reading
  • Accuracy established within the limits of error allowed by NIST™
Specifications
Specifications
0.84 oz.
Straight stopcock
±0.03mL
0.84 oz., 25mL
0.1mL
NIST
Glass
25mL
Schellbach Buret
Documents
Accessories