Please login to your online account to display your discounted pricing
Fisherbrand™ Schellbach Burets with Straight Std. Taper Glass Stopcock
$234.80 - $256.10
Specifications
Certifications/Compliance NIST
Product Type Schellbach Buret
Includes Straight stopcock
Stopcock Material Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity (English) Tolerance (Metric) Capacity (Metric) Subdivision Price Quantity    
03-740A
  Accessories
fisherbrand™
703740A
0.84 oz. ±0.03mL 25mL 0.1mL Each for $245.37
Case of 12 Each for $2,944.44
03-740B
  Accessories
fisherbrand™
703740B
1.69 oz. ±0.1mL 50mL 0.1mL Each for $234.80
Case of 12 Each for $2,817.60
03-740C
  Accessories
fisherbrand™
703740C
3.38 oz. ±0.2mL 100mL 0.2mL Each for $256.10
Case of 6 Each for $1,536.60
Description & Specifications
Specifications
Certifications/Compliance NIST
Product Type Schellbach Buret
Includes Straight stopcock
Stopcock Material Glass
  • Accentuate meniscus for easy reading
  • Accuracy established within the limits of error allowed by NIST™