Fisherbrand™ Sav-It™ Tube Closures
Firm, leak-resistant seals protect personnel and samples
Manufacturer:  Fisherbrand™ A21067FIS
 View more versions of this product
Catalog No. 02-707-14
Description
Description
  • One-hand application
  • Can be punctured for through-cap sampling via closed-tube instrumentation systems
  • Won't interfere with most common chemistry, coagulation, and drug monitoring methodologies
  • Fit test tubes, centrifuge tubes, specialty tubes, and evacuated blood drawing tubes
  • Direct through-cap sampling via closed-tube instrumentation systems
  • Won't crack or fall off
  • Hold fast under most rigorous procedures including agitation, centrifugation, refrigeration, and freezing
Specifications
Specifications
Non-sterile
Elastomer Co-polymer
Test tubes, centrifuge tubes, specialty tubes, evacuated blood drawing tubes
12 to 13mm
No
Gray
0.47 to 0.51 in.
Documents