Please login to your online account to display your discounted pricing

Fisherbrand™ Sav-It™ Tube Closures


Firm, leak-resistant seals protect personnel and samples

Manufacturer: fisherbrand™  A21070FIS

 View more versions of this product

Catalog No. 02-707-9

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Color Red
Sterile No
For Use With (Equipment) Test tubes, centrifuge tubes, specialty tubes, evacuated blood drawing tubes
Size (English) 0.47 to 0.51 in.
Size (Metric) 12 to 13mm
Autoclavable No
Material Elastomer Co-polymer