Please login to your online account to display your discounted pricing

Fisherbrand™ Sav-It™ Tube Closures


Firm, leak-resistant seals protect personnel and samples

Manufacturer: fisherbrand™  A21074FIS

 View more versions of this product

Catalog No. 02-707-5

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Color Blue
Sterile No
For Use With (Equipment) Test tubes, centrifuge tubes, specialty tubes, evacuated blood drawing tubes
Size (Metric) 16mm
Autoclavable No
Material Elastomer Co-polymer
Size (English) 0.62 in.