Please login to your online account to display your discounted pricing

Fisherbrand™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles


Eliminates drips and allows bottle to dispense upright or inverted, Bel Art™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles' color-coded polypropylene closures aid in quick and easy identification.

Manufacturer: fisherbrand™  118163011

 View more versions of this product

Catalog No. 08-647-712

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Closure Color Green
Autoclavable No
Mouth Wide
For Use With (Application) Methanol
Closure Material Polypropylene
Capacity (English) 16 oz.
Material LDPE
Color Green
Certifications/Compliance OSHA 29 CFR 1910.1200
Capacity (Metric) 500mL
Capacity 16 oz., 500mL

Preprinted bottles feature:

  • Keep liquids in clearly marked bottles that help prevent accidental misuse
  • Chemical name and formula
  • Full color NFPA™ Diamond
  • Health hazards
  • CAS # (Chemical Abstract Services #)
  • Suggested protective clothing and equipment
  • Wide mouth for easy filling
  • Color caps enhance identification
  • LDPE bottles with polypropylene valves
  • 16 oz. (500mL)
  • “Label Your Own” bottles also available
  • Sodium Hypochlorite (Bleach) labeled bottles are WHITE for light protection
  • Use only with solvents compatible with LDPE