missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles

Eliminates drips and allows bottle to dispense upright or inverted, Bel Art™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles' color-coded polypropylene closures aid in quick and easy identification.

Manufacturer:  Fisherbrand™ 118163019

 View more versions of this product

Catalog No. 08-647-710


 

Specifications

Description

Wash bottle, Plastic; Fisherbrand; Safety-Vented Right-to-Know; Pre-printed safety-labeled bottles; Wide-mouth for easy filling; LDPE with polypropylene cap/valve; Label: Ethanol; Cap: Natural; 16 oz. (500mL); 6/pk.

Preprinted bottles feature:

  • Keep liquids in clearly marked bottles that help prevent accidental misuse
  • Chemical name and formula
  • Full color NFPA™ Diamond
  • Health hazards
  • CAS # (Chemical Abstract Services #)
  • Suggested protective clothing and equipment
  • Wide mouth for easy filling
  • Color caps enhance identification
  • LDPE bottles with polypropylene valves
  • 16 oz. (500mL)
  • “Label Your Own” bottles also available
  • Sodium Hypochlorite (Bleach) labeled bottles are WHITE for light protection
  • Use only with solvents compatible with LDPE
Description & Specifications

Specifications

16 oz., 500mL
16 oz.
Polypropylene
Ethanol
No
Wide
500mL
OSHA 29 CFR 1910.1200
Natural
LDPE
Natural
Promotions

Promotions