Please login to your online account to display your discounted pricing

Fisherbrand™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles

Eliminates drips and allows bottle to dispense upright or inverted, Bel Art™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles' color-coded polypropylene closures aid in quick and easy identification.

$103.41 - $113.69

Specifications

Capacity 16 oz., 500mL
Certifications/Compliance OSHA 29 CFR 1910.1200
Closure Material Polypropylene
Autoclavable No
Material LDPE
View More Specs
Products
Catalog Number Mfr. No. Closure Color For Use With (Application) Color Price Quantity    

08-647-708

 
fisherbrand™
118163001
Red Acetone Red Pack of 6 Each for $103.41

08-647-707

 
fisherbrand™
118163050
Assorted Pack of 6 Each for $113.69

08-647-710

 
fisherbrand™
118163019
Natural Ethanol Natural Pack of 6 Each for $113.69

08-647-709

 
fisherbrand™
118163004
Blue Distilled Water Blue Pack of 6 Each for $110.10

08-647-712

 
fisherbrand™
118163011
Green Methanol Green Pack of 6 Each for $110.10

08-647-713

 
fisherbrand™
118163015
Yellow Sodium Hypochlorite Yellow Pack of 6 Each for $110.10

08-647-711

 
fisherbrand™
118163008
Yellow Isopropanol Yellow Pack of 6 Each for $110.10
Description & Specifications

Specifications

Capacity 16 oz., 500mL
Certifications/Compliance OSHA 29 CFR 1910.1200
Closure Material Polypropylene
Autoclavable No
Material LDPE
Mouth Wide

Preprinted bottles feature:

  • Keep liquids in clearly marked bottles that help prevent accidental misuse
  • Chemical name and formula
  • Full color NFPA™ Diamond
  • Health hazards
  • CAS # (Chemical Abstract Services #)
  • Suggested protective clothing and equipment
  • Wide mouth for easy filling
  • Color caps enhance identification
  • LDPE bottles with polypropylene valves
  • 16 oz. (500mL)
  • “Label Your Own” bottles also available
  • Sodium Hypochlorite (Bleach) labeled bottles are WHITE for light protection
  • Use only with solvents compatible with LDPE

Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.