missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Safety-Labeled Wash Bottles

Include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Bel Art™ Safety-Labeled Wash Bottles also include Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information to make your workplace safer.

Manufacturer:  Fisherbrand™ 006371502

 View more versions of this product

Catalog No. 03-409-23B


 

Specifications

Description

Bottle, Wash; Fisherbrand; Low-Density polyethlyene; Wide-mouth; Color-coded; Ethanol; Safety-Labeled DOT, NFPA, CAS protective euip and target organ; easy to read 17 oz. (500mL)

 

  • Easy-to-read printed bottles
  • Wide mouth 53mm (2") bottle for easy refilling
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Polypropylene closures
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Use only with solvents compatible with LDPE
Description & Specifications

Specifications

17 oz., 500mL
17 oz.
Ethanol
No
500mL
OSHA 29 CFR 1910.1200
Polyethylene
Wide
Accessories
Promotions

Promotions