Please login to your online account to display your discounted pricing

Fisherbrand™ Safety-Labeled Wash Bottles


Include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Bel Art™ Safety-Labeled Wash Bottles also include Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information to make your workplace safer.

Manufacturer: fisherbrand™  006221500

 View more versions of this product

Catalog No. 03-409-23A

  • / Pack of 6 Each
 

Description & Specifications

Specifications

For Use With (Application) Acetone
Mouth Wide
Autoclavable No
Capacity (English) 17 oz.
Certifications/Compliance OSHA 29 CFR 1910.1200
Material Polyethylene
Capacity 17 oz., 500mL
Capacity (Metric) 500mL

 

  • Easy-to-read printed bottles
  • Wide mouth 53mm (2") bottle for easy refilling
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Polypropylene closures
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Use only with solvents compatible with LDPE

Accessories

Accessories for Catalog Number: 0340923A

Item Mfr. No. Item Description Price Per Unit Quantity  
03-409-24
Bel-Art™ SP Scienceware™ Wash Bottle Carrier
bel-art™
F116400001
Transports up to 6 wash bottles. Bel Art™ SP Scienceware™ Wash Bottle Carrier offers safe, convenient, lightweight transportation that is sturdy and lightweight. Each for $159.14