Please login to your online account to display your discounted pricing

Fisherbrand™ Safety-Labeled Wash Bottles


Include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Bel Art™ Safety-Labeled Wash Bottles also include Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information to make your workplace safer.

Manufacturer: fisherbrand™  006121500

 View more versions of this product

Catalog No. 03-409-23R

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Capacity (English) 17 oz.
Certifications/Compliance OSHA 29 CFR 1910.1200
Material Polyethylene
Mouth Wide
Capacity (Metric) 500mL
Autoclavable No
For Use With Machine oil
Capacity 17 oz., 500mL
Accessories

Accessories for Catalog Number: 0340923R

Item Mfr. No. Item Description Price Per Unit Quantity