Please login to your online account to display your discounted pricing
Fisherbrand™ Safety-Labeled Wash Bottles
Include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Bel Art™ Safety-Labeled Wash Bottles also include Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information to make your workplace safer.
$50.67 - $58.17
Specifications
Certifications/Compliance OSHA 29 CFR 1910.1200
Autoclavable No
Material Polyethylene
Mouth Wide
View More Specs
Products
Catalog Number Mfr. No. Capacity For Use With Price Quantity    
03-409-23K
  Accessories
fisherbrand™
006201500
17 oz., 500mL Distilled Water Pack of 6 Each for $58.17
03-409-23Q
fisherbrand™
006221000
32 oz., 1000mL Acetone Pack of 6 Each for $57.84
03-409-23R
  Accessories
fisherbrand™
006121500
17 oz., 500mL Machine oil Pack of 6 Each for $50.67
03-409-23S
  Accessories
fisherbrand™
006111500
17 oz., 500mL Ethyl Ketone Pack of 6 Each for $50.67
03-409-23A
  Accessories
fisherbrand™
006221500
17 oz., 500mL Acetone Pack of 6 Each for $58.17
03-409-23D
  Accessories
fisherbrand™
006231500
17 oz., 500mL Methanol Pack of 6 Each for $58.17
03-409-23C
  Accessories
fisherbrand™
006241500
17 oz., 500mL Isopropanol Pack of 6 Each for $58.17
03-409-23B
  Accessories
fisherbrand™
006371502
17 oz., 500mL Ethanol Pack of 6 Each for $58.17
03-409-23H
  Accessories
fisherbrand™
006291502
17 oz., 500mL 6 Bottle variety pack that includes: Methanol, Isopropanol, Water, Acetone, Toluene, Ethanol Pack of 6 Each for $58.17
03-409-23G
  Accessories
fisherbrand™
006211500
17 oz., 500mL Water Pack of 6 Each for $53.79
03-409-23E
  Accessories
fisherbrand™
006281500
17 oz., 500mL Toluene Pack of 6 Each for $58.17
03-409-23J
  Accessories
fisherbrand™
006171500
17 oz., 500mL Saline Pack of 6 Each for $58.17
03-409-23L
  Accessories
fisherbrand™
006271500
17 oz., 500mL Bleach Pack of 6 Each for $58.17
03-409-23N
  Accessories
fisherbrand™
006381500
17 oz., 500mL Universal Pack of 6 Each for $58.17
03-409-23M
  Accessories
fisherbrand™
006311500
17 oz., 500mL Deionized Water Pack of 6 Each for $58.17
03-409-23P
  Accessories
fisherbrand™
006141500
17 oz., 500mL Soap Pack of 6 Each for $56.23
Description & Specifications
Specifications
Certifications/Compliance OSHA 29 CFR 1910.1200
Autoclavable No
Material Polyethylene
Mouth Wide
  • Easy-to-read printed bottles
  • Wide mouth 53mm (2") bottle for easy refilling
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Polypropylene closures
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Use only with solvents compatible with LDPE
Available in cases of six, all with the same label, or in a six-bottle assortment: one 500mL bottle each of acetone, ethanol, isopropanol, methanol, toluene and water.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.