Please login to your online account to display your discounted pricing

Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles


Keep liquids in clearly marked bottles that help prevent accidental misuse. Bel Art™ Safety-Labeled Right-to-Know Wash Bottles are available in both preprinted and “Label Your Own” bottles.

Manufacturer: fisherbrand™  117163001

 View more versions of this product

Catalog No. 08-647-701

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Closure Color Red
Mouth Wide
Autoclavable No
For Use With (Application) Acetone
Quantity 6/Pk.
Closure Material Polypropylene
Color Red
Capacity (English) 16 oz.
Certifications/Compliance OSHA 29 CFR 1910.1200
Material LDPE
Capacity (Metric) 500mL
Capacity 16 oz., 500mL

  • Chemical name and formula
  • Full color NFPA™ Diamond
  • Health hazards
  • CAS # (Chemical Abstract Services #)
  • Suggested protective clothing and equipment
  • Color caps enhance identification
  • LDPE bottles with polypropylene caps
  • “Label Your Own” bottles also available
  • Sodium Hypochlorite (Bleach) labeled bottles are white for light protection
  • Use only with solvents compatible with LDPE; Sodium Hypochlorite (Bleach) labeled bottles are WHITE for light protection