missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles

Keep liquids in clearly marked bottles that help prevent accidental misuse. Bel Art™ Safety-Labeled Right-to-Know Wash Bottles are available in both preprinted and “Label Your Own” bottles.

Manufacturer:  Fisherbrand™ 117163050

 View more versions of this product

Catalog No. 08-647-700


 

Specifications

Description

Wash bottle, Plastic; Fisherbrand; Right-to-Know; Pre-printed safety-labeled bottles; Feature chemical name and formula; Wide-mouth for easy filling; LDPE with polypropylene cap; Assortment pack; 16 oz. (500mL); 5/pk.

  • Chemical name and formula
  • Full color NFPA™ Diamond
  • Health hazards
  • CAS # (Chemical Abstract Services #)
  • Suggested protective clothing and equipment
  • Color caps enhance identification
  • LDPE bottles with polypropylene caps
  • “Label Your Own” bottles also available
  • Sodium Hypochlorite (Bleach) labeled bottles are white for light protection
  • Use only with solvents compatible with LDPE; Sodium Hypochlorite (Bleach) labeled bottles are WHITE for light protection
Description & Specifications

Specifications

16 oz., 500mL
16 oz.
Polypropylene
5/Pk.
No
Wide
500mL
OSHA 29 CFR 1910.1200
LDPE
Assortment Pack
Assorted
Promotions

Promotions