missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles

Keep liquids in clearly marked bottles that help prevent accidental misuse. Bel Art™ Safety-Labeled Right-to-Know Wash Bottles are available in both preprinted and “Label Your Own” bottles.

$44.76 - $64.52

Specifications

Material LDPE
Mouth Wide
Autoclavable No
Certifications/Compliance OSHA 29 CFR 1910.1200
Closure Material Polypropylene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Color For Use With (Application) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Closure Color For Use With (Application) Capacity (Metric) Price Quantity  
08-647-700 Fisherbrand™
117163050
Assorted 500mL Pack of 5 Each for $59.45
08-647-704 Fisherbrand™
117163008
Yellow Isopropanol 500mL Pack of 6 Each for $62.48
08-647-702 Fisherbrand™
117163004
Blue Distilled Water 500mL Pack of 6 Each for $62.48
08-647-701 Fisherbrand™
117163001
Red Acetone 500mL Pack of 6 Each for $62.48
08-647-705 Fisherbrand™
117163011
Green Methanol 500mL Pack of 6 Each for $62.48
08-647-706 Fisherbrand™
117163015
Yellow Sodium Hypochlorite 500mL Pack of 6 Each for $62.48
08-647-703 Fisherbrand™
117163019
Natural Ethanol 500mL Pack of 6 Each for $64.52
08-647-714 Fisherbrand™
117323004
Blue Distilled Water 1000mL Pack of 4 Each for $44.76
08-647-716 Fisherbrand™
117323008
Yellow Isopropanol 1000mL Pack of 4 Each for $44.76
08-647-715 Fisherbrand™
117323019
Natural Ethanol 1000mL Pack of 4 Each for $46.24
08-647-717 Fisherbrand™
117323011
Green Methanol 1000mL Pack of 4 Each for $44.76
08-647-718 Fisherbrand™
117323015
Yellow Sodium Hypochlorite 1000mL Pack of 4 Each for $44.76
Specifications

Description

  • Chemical name and formula
  • Full color NFPA™ Diamond
  • Health hazards
  • CAS # (Chemical Abstract Services #)
  • Suggested protective clothing and equipment
  • Color caps enhance identification
  • LDPE bottles with polypropylene caps
  • “Label Your Own” bottles also available
  • Sodium Hypochlorite (Bleach) labeled bottles are white for light protection
  • Use only with solvents compatible with LDPE; Sodium Hypochlorite (Bleach) labeled bottles are WHITE for light protection

Compliance

Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

LDPE
No
Polypropylene
Wide
OSHA 29 CFR 1910.1200