Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Coated Straight-Sided Jars with Polyvinyl-Lined Caps
$296.27 - $296.27
Specifications
Closure Liner Poly/Vinyl
Closure Material PTFE
Closure Screw
Closure Size 70-400
Color Clear
View More Specs
Products
Catalog Number Mfr. No. Capacity Price Quantity    
02-911-867
fisherbrand™
W014028
32 oz., 1000mL Case of 12 Each for $296.27
Description & Specifications
Specifications
Closure Liner Poly/Vinyl
Closure Material PTFE
Closure Screw
Closure Size 70-400
Color Clear
Material Polypropylene
Mouth Wide
  • Type III clear soda-lime glass with attached caps