Please login to your online account to display your discounted pricing
Fisherbrand™ Clear Boston Round Bottles with Black Phenolic Rubber Cap
Ideal lab bottle for handling volatile substances. Fisherbrand™ Clear Boston Round Bottles with Black Phenolic Rubber Cap are easier to grip and harder to break.
$125.07 - $125.07
Specifications
Closure Liner Rubber
Closure Material Phenolic
Closure Type Screw
Color Clear
Material Safety Coated Glass
View More Specs
Products
Catalog Number Mfr. No. Closure Size Capacity (English) Capacity (Metric) Price Quantity    
02-912-318
fisherbrand™
263881
28-400 16 oz. 480mL Case of 12 Each for $125.07
Description & Specifications
Specifications
Closure Liner Rubber
Closure Material Phenolic
Closure Type Screw
Color Clear
Material Safety Coated Glass
  • Boston Rounds that have been coated in a layer of Plastisol
  • If the glass does break, the plastisol coating contains substance in the bottle long enough to allow for proper disposal
  • Feature a round body, curved shoulders, and a narrow screw neck opening
  • Not autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.