Fisherbrand™ Safety Coated Clear Boston Round Bottles with White Polypropylene PE Foam/PTFE Cap
Ideal lab bottle for handling volatile substances. Fisherbrand™ Safety Coated Clear Boston Round Bottles with White Polypropylene Polypropylene PE Foam/PTFE Cap are easier to grip and harder to break.
$100.73 - $214.60
Specifications
Closure Included Yes
Closure Liner Polyethylene
Closure Material Polypropylene
Closure Type Screw
Color Clear
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Size Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Closure Size Capacity (English) Capacity (Metric) Price Quantity  
02-911-821 Fisherbrand™
263944
24-400 8 oz. 240mL Case of 12 Each for $100.73
02-911-822 Fisherbrand™
263882
28-400 16 oz. 480mL Case of 12 Each for $137.03
02-911-823 Fisherbrand™
263913
33-400 32oz. 960mL Case of 12 Each for $214.60
Specifications
Description
  • Plastic-coated Boston Round bottles are easier to grip and harder to break than ordinary glass bottles, yet still offer the chemical resistance and UV protection of amber glass
  • If the glass does break, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal
  • Boston Round bottles feature a round body, curved shoulders, and a narrow screw neck opening 
White Polypropylene Pulp/Vinyl Lined Caps
  • Offer good impact strength, thermal resistance and pliability
  • Autoclavable
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Yes
Polypropylene
Clear
Polyethylene
Screw
Safety Coated Glass