Please login to your online account to display your discounted pricing

Fisherbrand™ Safety Coated Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap


Ideal lab bottle for handling volatile substances. Fisherbrand™ Safety Coated Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap are easier to grip and harder to break.

Manufacturer: fisherbrand™  263883

 View more versions of this product

Catalog No. 02-911-819

  • / Case of 12 Each
 

Description & Specifications

Specifications

Closure Material Polypropylene
Product Type Safety Coated Boston Round Bottle
Capacity (Metric) 16 oz., 480mL
Mouth Narrow
Material Safety Coated Glass
Neck Finish 28-400
Closure Color White
Quantity 12/Cs.
Shape Boston Round
Liner Material Pulp/Vinyl
Closure Type Screw
Capacity (English) 16 oz.
Color Clear
Closure Screw
Closure Size 28-400
Capacity 16 oz., 500mL
Closure Liner Poly/Vinyl