Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Coated Clear Boston Round Bottles without Cap
Ideal lab bottle for handling volatile substances. Fisherbrand™ Safety Coated Clear Boston Round Bottles without Cap are easier to grip and harder to break.
$379.20 - $650.20
Specifications
Closure Type Screw
Color Clear
Material Safety Coated Glass
View More Specs
Products
Catalog Number Mfr. No. Closure Size Capacity (English) Capacity (Metric) Price Quantity    
02-911-922
  Accessories
fisherbrand™
263911
33-400 32oz. 960mL Case of 30 Each for $379.20
02-911-860
fisherbrand™
263941
24-400 8 oz. 240mL Case of 108 Each for $650.20
02-911-921
fisherbrand™
263879
28-400 16 oz. 480mL Case of 60 Each for $482.40
Description & Specifications
Specifications
Closure Type Screw
Color Clear
Material Safety Coated Glass
  • Coated in layer of plastisol
  • Shatter-resistant coating reduces chance of breakage
  • If broken, safety coating will contain contents long enough for disposal
  • Non-slip coating also allows greater ease of stability, wet or dry
  • Not autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.