Please login to your online account to display your discounted pricing
Fisherbrand™ Safety-Coated Amber Wide-Mouth Packers: No Caps
For field sampling applications
$78.17 - $78.17
Specifications
Closure Included Yes
Closure Type Attached cap
Closure Size 53-400
Color Amber
Material Safety Coated Glass
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Height (Metric) Capacity (Metric) Price Quantity    
02-911-830
fisherbrand™
264016
Case of 12 Each for $78.17
Description & Specifications
Specifications
Closure Included Yes
Closure Type Attached cap
Closure Size 53-400
Color Amber
Material Safety Coated Glass
  • Amber Type III soda-lime glass
  • Shatter-resistant coating reduces chance of breakage
  • If broken, safety coating contains contents long enough for disposal
  • Non-slip coating also allows greater ease of stability when wet or dry
  • Not autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.