Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles without Cap
General use bottles ideal when UV protection is important. Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles without Cap absorbs all wavelengths in the ultraviolet spectrum.
Manufacturer:  Fisherbrand™ 264007
 View more versions of this product
Catalog No. 02-912-317
Description
Description
Safety Coated Amber Wide Mouth Packer Bottles are normal glass packers coated in plastisol, which is a solution of PVC resin in a liquid plasticizer that cools to form a perfectly-fitting plastic coating. If the glass breaks, the plastic coating contains the substance in the bottle long enough to allow for proper disposal. Plus, these plastic coated glass packer bottles are harder to break and easier to grip than normal glass bottles, reducing the likelihood of breakage occurring in the first place. Amber Glass Packer Bottles feature a wide mouth and are a great general purpose bottle for solids. The amber color filters out ultraviolet wavelengths, the glass offers excellent chemical resistance with minimal chemical interference, and the plastisol coating offers additional protection against lab mishaps.
  • Features narrow screw neck opening, curved shoulders, and a round body
  • Offers minimal chemical interference with contents
  • Ensure integrity for long term storage and protection for UV-sensitive contents
Specifications
Specifications
8 oz., 250mL
8.5oz.
Safety Coated GlassþPlastic Coated Glass
Bottle, Safety Coated Wide Mouth PackerþWide Mouth Packer Bottle
45-400
45-400
Screw
250mL
AmberþAmber
24/Cs.
No
Wide
Round
Documents