Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Coated Amber Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Caps
Ideal lab bottles for handling volatile substances. Fisherbrand™ Safety Coated Amber Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Cap offer chemical resistance and UV protection.
$115.60 - $206.87
Specifications
Closure Included Yes
Closure Liner PTFE
Closure Material Polypropylene
Closure Type Screw
Color Amber
View More Specs
Products
Catalog Number Mfr. No. Closure Size Capacity (English) Capacity (Metric) Price Quantity    
02-911-814
fisherbrand™
263910
33-400 32oz. 960mL Case of 12 Each for $206.87
02-911-817
fisherbrand™
263878
28-400 16 oz. 480mL Case of 12 Each for $115.60
02-911-894
fisherbrand™
263927
22-400 4 oz. 120mL Case of 24 Each for $141.27
Description & Specifications
Specifications
Closure Included Yes
Closure Liner PTFE
Closure Material Polypropylene
Closure Type Screw
Color Amber
Material Safety Coated Glass
  • Plastic-coated Boston Round bottles are easier to grip and harder to break than ordinary glass bottles
  • If the glass does break, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal
  • Features a round body, curved shoulders, and a narrow screw neck opening
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.