missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Safety-Coated Amber Boston Rounds: With Polyvinyl-Lined Closures

Manufacturer:  Fisherbrand™ W014308

 View more versions of this product

Catalog No. 02-911-812


 

Specifications Description & Specifications

Specifications

16 oz., 500mL
16 oz.
Poly/Vinyl
Glass
Screw
500mL
PTFE
Amber
28-400