Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Coated Amber Boston Round Bottles with Black Phenolic PolyCone Cap
Ideal lab bottles for handling volatile substances. Fisherbrand™ Safety Coated Amber Boston Round Bottles with Black Phenolic PolyCone Cap offer chemical resistance and UV protection.
$101.70 - $205.90
Specifications
Closure Included Yes
Closure Liner Polyethylene
Closure Material Phenolic
Closure Type screw cap
Color Amber
View More Specs
Products
Catalog Number Mfr. No. Closure Size Capacity (English) Capacity (Metric) Price Quantity    
02-911-901
fisherbrand™
263909
33-400 32oz. 960mL Case of 12 Each for $205.90
02-911-969
fisherbrand™
263877
28-400 16 oz. 480mL Case of 12 Each for $116.27
02-911-896
fisherbrand™
263926
22-400mm 4 oz. 120mL Case of 24 Each for $140.63
02-911-970
fisherbrand™
263939
24-400 8 oz. 240mL Case of 12 Each for $101.70
Description & Specifications
Specifications
Closure Included Yes
Closure Liner Polyethylene
Closure Material Phenolic
Closure Type screw cap
Color Amber
Material Safety Coated Glass
  • Coated in a layer of Plastisol
  • Plastisol coating contains substance in bottle long enough to allow for proper disposal
  • Easier to grip and harder to break than ordinary glass bottles
  • Feature round body, curved shoulders, and a narrow screw neck opening
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.