missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Safety Coated Amber Boston Round Bottles with Black Phenolic PolyCone Cap

Ideal lab bottles for handling volatile substances. Fisherbrand™ Safety Coated Amber Boston Round Bottles with Black Phenolic PolyCone Cap offer chemical resistance and UV protection.

Manufacturer:  Fisherbrand™ 263877

 View more versions of this product

Catalog No. 02-911-969



Description

Description

Safety Coated Amber Glass Boston Round Bottles are glass Boston Rounds that have been coated in a layer of Plastisol. These plastic-coated Boston Round bottles are easier to grip and harder to break than ordinary glass bottles, yet still offer the chemical resistance and UV protection of amber glass. If the glass does break, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal. Boston Round bottles feature a round body, curved shoulders, and a narrow screw neck opening. Plastic-coated Boston Rounds are ideal lab bottles for handling volatile substances.

  • Coated in a layer of Plastisol
  • Plastisol coating contains substance in bottle long enough to allow for proper disposal
  • Easier to grip and harder to break than ordinary glass bottles
  • Features a round body, curved shoulders, and a narrow screw neck opening

Specifications

Specifications

16 oz., 500mL
16 oz.
Polyethylene
Safety Coated Glass, Plastic Coated Glass
Bottle, Safety Coated Boston Round, Boston Round Bottle
28-400
Narrow
Boston Round
480mL
Phenolic
Amber, Amber
12/Cs.
Black
PolyCone
28-400
Screw
Documents
Promotions

Promotions