Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Blue Thermohydrometer, Fahrenheit
Manufacturer: fisherbrand™  1801SB
 View more versions of this product
Catalog No. 11-555-234
  • / Each
Description & Specifications
Specifications
ASTM Number 6H
Certifications/Compliance NIST
Ballast Type Lead and mercury-free
Specific Gravity Range 10 to 30