Fisherbrand™ Safety Blue Thermohydrometer, Fahrenheit
Lead and mercury free
Manufacturer:  Fisherbrand™ 1984SB
 View more versions of this product
Catalog No. 11-555-206
Specifications
Description
Thermohydrometer, Safety Blue; Fisherbrand; Lead and mercury free; Temp. range: 30 to 150degF; Specific gravity: 1.600 to 1.820
  • Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Description & Specifications
Specifications
Lead and mercury-free
NIST
1.600 to 1.820
Thermohydrometer