Fisherbrand™ Safety Blue Thermohydrometer, Fahrenheit
Lead and mercury free
Manufacturer:  Fisherbrand™ 1912SB
 View more versions of this product
Catalog No. 11-555-202
Specifications
Description
Thermohydrometer, Safety Blue; Fisherbrand; Lead and mercury free; Temp. range: 0 to 150degF; Specific gravity: 0.900 to 1.010
  • Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Description & Specifications
Specifications
Lead and mercury-free
NIST
0.9 to 1.01
Thermohydrometer