Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Blue Thermohydrometer, Fahrenheit
Lead and mercury free
Manufacturer: fisherbrand™  1911SB
 View more versions of this product
Catalog No. 11-555-201
  • / Each
Description & Specifications
Specifications
Ballast Type Lead and mercury-free
Certifications/Compliance NIST
Specific Gravity Range 0.800 to 0.910
Product Type Thermohydrometer
  • Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.