Fisherbrand™ Safety Blue Thermohydrometer, Celsius
Lead and mercury free
Manufacturer:  Fisherbrand™ 8008SB
 View more versions of this product
Catalog No. 11-555-216
Specifications
Description
Thermohydrometer, Safety Blue; Fisherbrand; Lead and mercury free; Temp. range: 0 to 50deg.C; Brix: 69 to 81
  • Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Description & Specifications
Specifications
Lead and mercury-free
For determining the percentage by weight of sucrose
Thermohydrometer
NIST
69 to 81