Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Blue Thermohydrometer, Celsius
Manufacturer: fisherbrand™  8006SB
 View more versions of this product
Catalog No. 11-555-214
  • / Each
Description & Specifications
Specifications
For Use With (Application) For determining the percentage by weight of sucrose
Brix Range 49 to 61
Ballast Type Lead and mercury-free
Certifications/Compliance NIST