Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Blue Thermohydrometer, Celsius
Manufacturer: fisherbrand™  8002SB
 View more versions of this product
Catalog No. 11-555-210
  • / Each
Description & Specifications
Specifications
Ballast Type Lead and mercury-free
For Use With (Application) For determining the percentage by weight of sucrose
Brix Range 9 to 21
Certifications/Compliance NIST