Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Blue Thermohydrometer, Celsius
Manufacturer: fisherbrand™  8013SB
 View more versions of this product
Catalog No. 11-555-208
  • / Each
Description & Specifications
Specifications
For Use With (Application) For determining the percentage by weight of sucrose
Ballast Type Lead and mercury-free
Brix Range -5 to 5
Certifications/Compliance NIST