Fisherbrand™ Safety Blue Baume (Heavy) Thermohydrometers
Lead and mercury free
Manufacturer:  Fisherbrand™ 1931SB
 View more versions of this product
Catalog No. 11-555-239
Specifications
Description
Thermohydrometer, Safety Blue; Fisherbrand; Lead and mercury free; Temp. range: 20 to 150degF; Baume: 9 to 21
  • Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Description & Specifications
Specifications
Lead and mercury-free
NIST
Baume: 9 to 21
Thermohydrometer