Fisherbrand™ Safety Blue Baume (Heavy) Thermohydrometers
Lead and mercury free
Manufacturer:  Fisherbrand™ 1939SB
 View more versions of this product
Catalog No. 11-555-246
Specifications
Description
Thermohydrometer, Safety Blue; Fisherbrand; Lead and mercury free; Temp. range: 30 to 150degF; Baume: 35 to 70
  • Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Description & Specifications
Specifications
Lead and mercury-free
NIST
Baume: 35 to 70
Thermohydrometer