Please login to your online account to display your discounted pricing

Fisherbrand™ Black Plastic Bottle Screw Caps


Manufacturer: fisherbrand™  W014549

 View more versions of this product

Catalog No. 02-912-051

  • / Case of 576 Each
 

Description & Specifications

Specifications

Liner Material Rubber
Closure Size 33/400
Closure Type Screw cap
Sterility Sterile
Autoclavable Yes
Neck Finish 33-400
For Use With (Equipment) Bottle
Closure Material Plastic
Description Screw cap
Quantity 576/PK.
Length (Metric) 36mm
Material Plastic
Certifications/Compliance ISO 9001
Color Black
No. per Case 576 Cs.
Closure Liner 14B White Rubber