Please login to your online account to display your discounted pricing

Fisherbrand™ Black Plastic Bottle Screw Caps


Manufacturer: fisherbrand™  W014548

 View more versions of this product

Catalog No. 02-912-050

  • / Case of 576 Each
 

Description & Specifications

Specifications

Autoclavable Yes
Liner Material Rubber
Sterility Sterile
Closure Type Screw cap
For Use With (Equipment) Bottle
Closure Size 28/400
Neck Finish 28-400
Closure Material Plastic
Length (Metric) 80mm
Description Screw cap
Quantity 576/PK.
Certifications/Compliance ISO 9001
Material Plastic
Color Black
Closure Liner 14B White Rubber
No. per Case 576 Cs.