Fisherbrand™ Rubber Bulb-Type Safety Pipet Fillers
Tough 100% natural rubber patented pipet fillers eliminate mouth pipetting and handling of hazardous materials
Manufacturer:  Fisherbrand™ JUI1368151
 View more versions of this product
Catalog No. 13-681-51
Description
Description
Tough 100% natural rubber patented pipet fillers eliminate mouth pipetting and handling of hazardous materials
Two stainless-steel ball valves control vacuum for precise pipet filling and liquid delivery with one hand.
  • Expel air by squeezing bulb, then draw up liquid by pressing valve “↑” on stem
  • To release, press valve “↓” on side of stem
  • Fit pipets with O.D. ranging from 0.187 to 0.375 in. (4.5 to 9mm)
  • Flip style pipette fillers enables one handed bulb operation and removal of top white cap to clean out the bulb from inside
Specifications
Specifications
No
Two stainless steel ball valves
Red
Pipet
Natural Rubber
Pipet Filler
Documents