Dynalon™ Round Dipping Baskets
Sturdy and chemical resistant.
Supplier:  Dynalon™ 146604
 View more versions of this product
Catalog No. 14-965-2F
Description
Description
  • HDPE fabricated dipping baskets are made with 1/8 in. thick walls, 3/16 in. diameter perforations on 5/16 in. centers
  • Made from high density polyethylene
  • Takes 140°F on a continuous basis or more for shorter periods (up to 160°F)
  • Supplied with sturdy swinging handle—a unique feature allowing easy storage and loading
  • All have perforated bottoms
  • Polypropylene fabricated dipping baskets withstands 180°F on a continuous basis or up to 200°F for shorter periods
  • Excellent chemical resistance
  • Supplied with swinging handle
  • Perforations 1/8 in. diameter on 21/64 in. center
Order Info
Additional dipping basket sizes available. To order, contact your Fisher Scientific Customer Service Representative.
Specifications
Specifications
Dipping Basket
White
1
6 x 6 in.
16.827 cm
16.192 cm
21.907 cm
15.24 cm
Autoclavable
Polypropylene
Non-sterile
6.625 x 6.375 x 8.625 in. (16.827 x 16.192 x 21.907cm)
6.625 in.
6.375 in.
8.625 in.