missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Round Covered Lead Flask Rings
Stabilize Erlenmeyer flasks immersed in water baths.
Manufacturer: Fisherbrand™ 183073005
Description
Stabilize Erlenmeyer flasks immersed in water baths.
- Reduces floating, breakage and chemical spills
- Bright blue coating
- Chemical-resistant
- Will not scratch flasks
- Available in four diameter/weight combinations
Specifications
4.1 cm | |
Blue | |
125 to 500mL Erlenmeyer Flasks Immersed in Water Baths | |
Lead | |
226.7 g |
Not Autoclavable | |
1.6 in. | |
Flasks (125 to 500 mL) | |
0.5 lb. | |
Accessory |