Please login to your online account to display your discounted pricing

Fisherbrand™ Robotic Screw Top Autosampler Vial Closures


For use with Fisherbrand Robotic Screw Top Autosampler Vials (cat. no. 033918)

Manufacturer: fisherbrand™  0339132

 View more versions of this product

Catalog No. 03-391-32

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Closure Type Screw top
Material PTFE/Silicone/PTFE
Color Blue
Quantity 100/Pk.
Type Closure
Vial Closure Size 9mm