missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ RNase Displace™ Decontaminant

Removes RNase and DNA contamination without a time-consuming autoclaving step

$67.17 - $256.80

Specifications

Boiling Point >212°F
Color Milky White
Composition <1% Potassium Hydroxide
For Use With (Application) All surfaces, including Glass, Plastic, Metal and Ceramic, removes DNase and DNA contamination without a time-consuming autoclaving step
Format Liquid
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
04-355-136 View Documents Fisherbrand™
1401**
1 gal. Each for $256.80
Case of 4 Each for $1,027.20
04-355-137 View Documents Fisherbrand™
1408
8 oz. Each for $67.17
Case of 12 Each for $806.04
04-355-138 View Documents Fisherbrand™
1432
32 oz. Each for $121.27
Case of 12 Each for $1,455.24
Description

Description

 

  • Can be used on all surfaces, including glass, plastic, metal, and ceramic
  • Works instantly
  • Totally rinsable from the decontaminated surface
  • Phosphate-free

 

Specifications

Specifications

>212°F
<1% Potassium Hydroxide
Liquid
2 to 12 hr.
Milky White
All surfaces, including Glass, Plastic, Metal and Ceramic, removes DNase and DNA contamination without a time-consuming autoclaving step
13