Please login to your online account to display your discounted pricing

Fisherbrand™ RNase Displace™ Decontaminant


Removes RNase and DNA contamination without a time-consuming autoclaving step

Manufacturer: fisher scientific  1432

 View more versions of this product

Catalog No. 04-355-138

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Color Milky White
Test Time 2 to 12 hr.
For Use With (Application) All surfaces, including Glass, Plastic, Metal and Ceramic, removes DNase and DNA contamination without a time-consuming autoclaving step
pH Range 13
Boiling Point >212°F
Composition <1% Potassium Hydroxide
Format Liquid
Size 32 oz.