Fisherbrand™ RNase Displace™ Decontaminant
Removes RNase and DNA contamination without a time-consuming autoclaving step
Manufacturer:  Fisher Scientific 1408
 View more versions of this product
Catalog No. 04-355-137
Specifications Description & Specifications
Specifications
All surfaces, including Glass, Plastic, Metal and Ceramic, removes DNase and DNA contamination without a time-consuming autoclaving step
Liquid
2 to 12 hr.
8 oz.
>212°F
<1% Potassium Hydroxide
Milky White
13