Fisherbrand™ Reusable Glass Tubes with Phenolic Screw Caps
Phenolic screw cap with white rubber liner
Manufacturer:  Fisherbrand™ K73750M96
 View more versions of this product
Catalog No. 14-959-25E
Description
Description
Phenolic screw cap with white rubber liner
  • High-quality borosilicate glass
  • Marking spot
Order Info
Packed in shrink-wrapped, lint-free trays
Specifications
Specifications
Phenolic
Suitable for tissue culture work and general bacteriological use
Rubber
25mL
125mm
Disposable Culture Tube
Non-sterile
20mm
Marking Spot
18-415
Screw
Borosilicate Glass
Round Bottom
Documents
Accessories