Please login to your online account to display your discounted pricing

Fisherbrand™ Reusable Glass Class A Volumetric Flasks with standard taper Stopper


Single graduation ring blasted on the neck

Manufacturer: fisherbrand™  FS2801450

 View more versions of this product

Catalog No. 10-205B

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Calibration To contain
Stopper Material Glass
Total Count 1.0
Tolerance ±0.05mL
Stopper Number 9
Capacity (Metric) 50mL
Graduated Yes
Material Glass
Accessories

Accessories for Catalog Number: 10205B

Item Mfr. No. Item Description Price Per Unit Quantity  
04-320-4
Fisherbrand™ Sparkleen™ 1 Detergent
fisherbrand™
043204
Removes proteins, fats, oils, media, wax pencil marks, dried blood and other organic-inorganic deposits from glassware Each for $17.76
Case of 12 Each for $213.12