Please login to your online account to display your discounted pricing

Fisherbrand™ Reusable Glass Class A Volumetric Flasks with Snap Caps


Class A accuracy–without a serial number

Manufacturer: fisherbrand™  FS28008100

 View more versions of this product

Catalog No. 10-200C

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Stopper Material Polyethylene
Total Count 1.0
Tolerance ±0.08mL
Material Glass
Calibration To contain
Capacity (Metric) 100mL
Stopper Number 16
Certifications/Compliance Class A, ASTM E288
Graduated Yes
Accessories

Accessories for Catalog Number: 10200C

Item Mfr. No. Item Description Price Per Unit Quantity  
04-320-4
Fisherbrand™ Sparkleen™ 1 Detergent
fisherbrand™
043204
Removes proteins, fats, oils, media, wax pencil marks, dried blood and other organic-inorganic deposits from glassware Each for $17.76
Case of 12 Each for $213.12