Fisherbrand™ Replacement Red-Spirit™ Pocket Thermometers
Pocket thermometers
Manufacturer:  HB Instrument 608703420
 View more versions of this product
Catalog No. 15-021-5C
Specifications
Specifications
−30° to 120°F
−30°F
5.70 in.
120°F
Total Immersion
145mm
Documents